Furan-2,5-dicarboxylic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,5-Furandicarboxylic acid
H28718.03 H28718.06
H28718.03EA 2050 SEK
H28718.03 H28718.06
Furan-2,5-dicarboxylic acid ≥98%
Furan-2,5-dicarboxylic acid
Purity: 98% CAS No.: 3238-40-2 Molecular Formula: C6H4O5 Formula Weight: 156.09 Melting Point: 310° MDL No.: MFCD00016582
Formel: C₆H₄O₅
MW: 156,09 g/mol
Smältpunkt: >310 °C
Storage Temperature: Omgivande
MDL Number: MFCD00016582
CAS nummer: 3238-40-2
EINECS: 221-800-8