Printers with laboratory information label kit
Leverantör: Brady
710900 710898 710933 710942
BRDY710900EA 7600 SEK
BRDY710900 116-0374 116-0349 116-0351
Printers with laboratory information label kit
Printer Label Printers
These kits provide the user with printer and a selection of labels to accurately identify samples in the laboratory. Both the BMP™51 and BMP™53 are portable, single colour, thermal printers with 300 dpi print head and with very similar specifications, except the BMP™51 has stand alone capability and the BMP™53 does not. The top-loading label cartridge contains both the printer ribbon and labels and can be exchanged in seconds.
  • USB, Bluetooth and optional Ethernet, WiFi connectivity
  • Print speed 25,4 mm/second
  • BMP™51 Laboratory identification kit has
More Product Information