Capillary columns, CP-FFAP CB
Leverantör: VARIAN
CP7485 CP7485I5 CP7486 CP7686
568-0698EA 21950 SEK
568-0698 568-0699 568-0700 568-0715
Capillary columns, CP-FFAP CB
Kromatografikolonn
Columns for free fatty acids in dairy products.
  • Ideal for flavors, aromas and free fatty acids C₁ to C₂₆
  • Separates C₂ to C₂₄ acids in one run without derivatisation, saving time
  • Water and solvent resistant
  • Chemically-bonded
Temperature limits: 250/275 °C.