Gel permeation chromatography columns, PolyPore™
Leverantör: VARIAN
PL1113-1500 PL1113-6500 PL1513-1500
VARIPL1113-1500EA 15425 SEK
VARIPL1113-1500 VARIPL1113-6500 VARIPL1513-1500
Gel permeation chromatography columns, PolyPore™
Kromatografikolonn
PolyPore columns results unrivalled resolution for the analysis of polymers with broad molecular weight distributions.
  • Routine polymer analysis with very high resolution
  • Wide operating range simplifies column choice
  • Low particle size extracts maximum information from the analyte