HPLC columns, NUCLEOSIL® SiOH
Leverantör: MACHEREY-NAGEL
720093.46 720099.46 721518.30 721167.30
MANA720093.46EA 11950 SEK
MANA720093.46 554-5799 554-5804 MANA721167.30
HPLC columns, NUCLEOSIL® SiOH
Kromatografikolonn
NUCLEOSIL® SiOH is an unmodified silica phase for HPLC.
  • Robust spherical silica gel
  • Very polar phase for NP chromatography
  • Separation mechanism based on polar interactions
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).