Allylboronic acid pinacol ester ≥96.0% stabilised
Leverantör: TCI
|
Varning
|
Synonyms:
2-Allyl-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane
Stabilization:
Stabilised with phenothiazine
A2157-1G
A2157-5G
TCIAA2157-1GEA
335
SEK
TCIAA2157-1G
TCIAA2157-5G
Allylboronic acid pinacol ester ≥96.0% stabilised
Allylboronic acid pinacol ester
Formel:
C₉H₁₇BO₂ Kokpunkt: 35°C Flampunkt: 46°C Storage Temperature: Frys |
MDL Number:
MFCD00013347 CAS nummer: 72824-04-5 UN: 3272 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...