Propylene glycol diacetate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Propane-1,2-diyl diacetate, 1,2-Propylene glycol diacetate, 1,2-Propanediol diacetate
P0902-25ML P0902-500ML
TCIAP0902-25MLEA 215 SEK
TCIAP0902-25ML TCIAP0902-500ML
Propylene glycol diacetate ≥98.0%
Propylene glycol diacetate
Formel: C₇H₁₂O₄
MW: 160,1678 g/mol
Kokpunkt: 186°C
Smältpunkt: -31°C
Flampunkt: 89°C
MDL Number: MFCD00026201
CAS nummer: 623-84-7
EINECS: 210-817-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=125a653de4b6b152ec30fab5cb7b1514ed3fbf5a&vid=125a653de4b6b152ec30fab5cb7b1514ed3fbf5a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %