4-Cumylphenol ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-(2-Phenylpropan-2-yl)phenol
C0684-25G
C0684-500G
TCIAC0684-25GEA
215
SEK
TCIAC0684-25G
TCIAC0684-500G
4-Cumylphenol ≥98.0%
4-Cumylphenol
Formel:
C₁₅H₁₆O MW: 212,29 g/mol |
MDL Number:
MFCD00002365 CAS nummer: 599-64-4 EINECS: 209-968-0 UN: 3077 ADR: 9,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...