(Trifluoromethyl)trimethylsilane 0.5 M i THF
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: trimethyl(trifluoromethyl)silane
H31520.AA H31520.AC H31520.AE
H31520.AAEA 580 SEK
H31520.AA H31520.AC H31520.AE
(Trifluoromethyl)trimethylsilane 0.5 M i THF
(Trifluoromethyl)trimethylsilane
CAS No.: 81290-20-2 Molecular Formula: C4H9F3Si Formula Weight: 142.20 Flash Point: 17°(63°F) Boiling Point: 40° UN No.: 1993 MDL No.: MFCD00145454
Formel: C₄H₉F₃Si
MW: 142.2 g/mol
Kokpunkt: 40 °C
Densitet: 0.895
Flampunkt: 17 °C (63 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00145454
CAS nummer: 81290-20-2
UN: 1993
ADR: 3 II