Aluminium tri-sec-butanolate 98,0 Titrimetrisk

Leverantör: TCI
Fara

Synonyms: Aluminium Tri-sec-butylate, Aluminum s-butoxide, Aluminium tri-sec-butoxide

A0244-25G A0244-500G
TCIAA0244-25GEA 230 SEK
TCIAA0244-25G TCIAA0244-500G
Aluminium tri-sec-butanolate 98,0 Titrimetrisk
Aluminium tri-sec-butanolate
Formel: C₁₂H₂₇AlO₃
MW: 246.33 g/mol
Flampunkt: 26°C
MDL Number: MFCD00009327
CAS nummer: 2269-22-9
EINECS: 218-871-2
UN: 1992
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc593fd0731a5ccb8a61f2a98ebab8e3f396c53c&vid=bc593fd0731a5ccb8a61f2a98ebab8e3f396c53c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Yellow to Green clear liquid
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR