Phosphoryl tribromide 95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Phosphorus(V) oxybromide, Phosphorus (V) oxide bromide, Phosphoryl bromide, Phosphorus oxybromide
388450250 388450100 388451000
ACRO388450250EA 3175 SEK
ACRO388450250 ACRO388450100 ACRO388451000
Phosphoryl tribromide 95%
Phosphoryl tribromide
Formel: Br₃OP
MW: 286.69 g/mol
Kokpunkt: 192 °C (760 mmHg)
Smältpunkt: 56 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00036298
CAS nummer: 7789-59-5
EINECS: 232-177-7
UN: 1939
ADR: 8 II
Merck Index: 12,07505
Order Now
Specifikation testresultat
Appearance Colorless to grey-brown Crystals or fused mass
Titration Argentometric ≥94 %