1-(1-Cyclopenten-1-yl)pyrrolidine ≥97.0%
Leverantör: TCI
|
Varning
|
Synonyms:
1-Pyrrolidino-1-cyclopentene
P1001-25G
P1001-5G
TCIAP1001-25GEA
3350
SEK
TCIAP1001-25G
TCIAP1001-5G
1-(1-Cyclopenten-1-yl)pyrrolidine ≥97.0%
1-(1-Cyclopenten-1-yl)pyrrolidine
Formel:
C₉H₁₅N Kokpunkt: 93°C Flampunkt: 47°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00003162 CAS nummer: 7148-07-4 EINECS: 230-463-6 UN: 1993 ADR: 3,III |
|
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...