Cyclohexanemethylamine ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
(Cyclohexylmethyl)amine, (Aminomethyl)cyclohexane, Cyclohexylmethanamine
A1114-100ML
A1114-25ML
TCIAA1114-100MLEA
2750
SEK
TCIAA1114-100ML
TCIAA1114-25ML
Cyclohexanemethylamine ≥98.0%
Cyclohexanemethylamine
Formel:
C₇H₁₅N MW: 113.2 g/mol Kokpunkt: 163°C Smältpunkt: -8 Flampunkt: 48°C Storage Temperature: Omgivande |
MDL Number:
MFCD00001520 CAS nummer: 3218-02-8 EINECS: 221-741-8 UN: 2734 ADR: 8 II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...