4,4'-Azoxyphenetole ≥98.0%

Leverantör: TCI

A0683-1G A0683-5G
TCIAA0683-1GEA 390 SEK
TCIAA0683-1G TCIAA0683-5G
4,4'-Azoxyphenetole ≥98.0%
4,4'-Azoxyphenetole
Formel: C₁₆H₁₈N₂O₃
MDL Number: MFCD00026790
CAS nummer: 4792-83-0
EINECS: 225-347-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9eb97b07cd079d039cbd690d7f0acd487d8611d&vid=d9eb97b07cd079d039cbd690d7f0acd487d8611d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR