Vinyl benzoate ≥99.0% stabilised
Leverantör: TCI
|
Varning
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
B1032-25ML
B1032-500ML
TCIAB1032-25MLEA
285
SEK
TCIAB1032-25ML
TCIAB1032-500ML
Vinyl benzoate ≥99.0% stabilised
Vinyl benzoate
Formel:
C₉H₈O₂ Kokpunkt: 203°C Flampunkt: 82°C |
MDL Number:
MFCD00048141 CAS nummer: 769-78-8 EINECS: 212-214-3 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...