Pyridine-2-carbaldehyde ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 2-Pyridinecarboxaldehyde, 2-Pyridinecarbaldehyde, Pyridine-2-carboxaldehyde
P0425-25G P0425-500G P0425-100G
TCIAP0425-25GEA 685 SEK
TCIAP0425-25G TCIAP0425-500G TCIAP0425-100G
Pyridine-2-carbaldehyde ≥98.0%
Pyridine-2-carbaldehyde
Formel: C₆H₅NO
MW: 107,11 g/mol
Kokpunkt: 181°C
Flampunkt: 77°C
Storage Temperature: Kylskåp
MDL Number: MFCD00006290
CAS nummer: 1121-60-4
EINECS: 214-333-6
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bac6f020cb5a4d0f36e3fb00422f276955db31b2&vid=bac6f020cb5a4d0f36e3fb00422f276955db31b2&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %