2-(Trimethylsilyl)thiazole ≥95.0%
Leverantör: TCI
Varning
T1299-1ML T1299-5ML
TCIAT1299-1MLEA 470 SEK
TCIAT1299-1ML TCIAT1299-5ML
2-(Trimethylsilyl)thiazole ≥95.0%
2-(Trimethylsilyl)thiazole
Formel: C₆H₁₁NSSi
Kokpunkt: 57°C
Flampunkt: 62°C
Storage Temperature: Kylskåp
MDL Number: MFCD00066274
CAS nummer: 79265-30-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e56a4e2dc949f79b4f9d7dc77a1ed72ec0e41c8&vid=2e56a4e2dc949f79b4f9d7dc77a1ed72ec0e41c8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %