Nerol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: cis-3,7-Dimethyl-2,6-octadien-1-ol

N0077-25ML N0077-500ML
TCIAN0077-25MLEA 230 SEK
TCIAN0077-25ML TCIAN0077-500ML
Nerol ≥98.0%
Nerol
Formel: C₁₀H₁₈O
MW: 154,25 g/mol
Kokpunkt: 226°C
Flampunkt: 93°C
MDL Number: MFCD00063204
CAS nummer: 106-25-2
EINECS: 203-378-7
Merck Index: 14,06475
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98bca1b24d1fb185d506535701061f5b1b5b308e&vid=98bca1b24d1fb185d506535701061f5b1b5b308e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR