1-Ethyl-3-methylimidazolium iodide ≥97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-ethyl-3-methylimidazolium iodide
H59404.06 H59404.18
H59404.06EA 875 SEK
H59404.06 H59404.18
1-Ethyl-3-methylimidazolium iodide ≥97%
1-Ethyl-3-methylimidazolium iodide
Formel: C₆H₁₁IN₂
Storage Temperature: Omgivande
MDL Number: MFCD03701101
CAS nummer: 35935-34-3