1-Chloro-3-iodobenzene ≥97.0% stabilised

Leverantör: TCI
Varning


Stabilization: Stabilised with copper chip

C0190-25G C0190-5G
TCIAC0190-25GEA 895 SEK
TCIAC0190-25G TCIAC0190-5G
1-Chloro-3-iodobenzene ≥97.0% stabilised
1-Chloro-3-iodobenzene
Formel: C₆H₄ClI
Kokpunkt: 228°C
Smältpunkt: 54,5 °C
Densitet: 1,9255 g/cm³ (20 °C)
Flampunkt: 101°C
MDL Number: MFCD00001046
CAS nummer: 625-99-0
EINECS: 210-920-6

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR