4-Hexyloxybenzoic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-(Hexyloxy)benzoic acid

H0247-25G H0247-500G
TCIAH0247-25GEA 605 SEK
TCIAH0247-25G TCIAH0247-500G
4-Hexyloxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Hexyloxybenzoic acid
Formel: C₁₃H₁₈O₃
MW: 222,28 g/mol
Smältpunkt: 150…153 °C
Densitet: ~ 1 g/cm³ (20 °C)
MDL Number: MFCD00013991
CAS nummer: 1142-39-8
EINECS: 214-534-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f54543b7681f0f912e84da135baff40c5ef6e32c&vid=95d5c507690e4a6f52ea04d11e4c2bbc5eb8b566&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR