4-Heptyloxybenzoic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
4-(Heptyloxy)benzoic acid
H0248-25G
H0248-500G
TCIAH0248-25GEA
735
SEK
TCIAH0248-25G
TCIAH0248-500G
4-Heptyloxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Heptyloxybenzoic acid
Formel:
C₁₄H₂₀O₃ Kokpunkt: 362 °C (1013 hPa) Smältpunkt: 146…146 °C Densitet: 1,183 g/cm³ (20 °C) |
MDL Number:
MFCD00013992 CAS nummer: 15872-42-1 EINECS: 239-995-3 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Mesomorphic range | 91.0 to 150.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...