2-Hydroxy-3-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-(Trifluoromethyl)pyridin-2-ol
T2600-1G
T2600-5G
TCIAT2600-1GEA
725
SEK
TCIAT2600-1G
TCIAT2600-5G
2-Hydroxy-3-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
2-Hydroxy-3-(trifluoromethyl)pyridine
Formel:
C₆H₄F₃NO MW: 163,1 g/mol |
MDL Number:
MFCD00153194 CAS nummer: 22245-83-6 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 153.0 to 157.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...