2,2,2-Trifluoroethyl methacrylate ≥98.0% stabilised
Leverantör: TCI EUROPE
Varning
Synonyms: Methacrylic acid 2,2,2-trifluoroethyl ester
Stabilization: Stabilised with 4-methoxyphenol (MEHQ)
M0738-25ML M0738-500ML M0738-100ML
TCIAM0738-25MLEA 245 SEK
TCIAM0738-25ML TCIAM0738-500ML TCIAM0738-100ML
2,2,2-Trifluoroethyl methacrylate ≥98.0% stabilised
2,2,2-Trifluoroethyl methacrylate
Formel: C₆H₇F₃O₂
MW: 168,12 g/mol
Kokpunkt: 102°C
Flampunkt: 25°C
MDL Number: MFCD00013576
CAS nummer: 352-87-4
EINECS: 206-525-3
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f701dbecb6eaccaad953e95fb28be79399ac8f99&vid=f701dbecb6eaccaad953e95fb28be79399ac8f99&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %