1-Bromo-3-methyl-2-butene ≥90.0% stabilised
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3,3-Dimethylallyl bromide, 4-Bromo-2-methyl-2-butene
Stabilization:
Stabilised with silver chip
B0844-25G
B0844-5G
TCIAB0844-25GEA
1050
SEK
TCIAB0844-25G
TCIAB0844-5G
1-Bromo-3-methyl-2-butene ≥90.0% stabilised
1-Bromo-3-methyl-2-butene
Formel:
C₅H₉Br MW: 149,03 g/mol Kokpunkt: 135°C Densitet: 1,293 g/cm³ (15 °C) Flampunkt: 40°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00000242 CAS nummer: 870-63-3 EINECS: 212-799-5 UN: 2920 ADR: 8,II |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...