(S)-(+)-Epiklorhydrin ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
(S)-(+)-3-Klor-1,2-epoxipropan, (S)-(+)-1-Klor-2,3-epoxipropan
E0533-25G
E0533-5G
TCIAE0533-25GEA
1225
SEK
TCIAE0533-25G
TCIAE0533-5G
(S)-(+)-Epiklorhydrin ≥98.0%
(S)-(+)-Epiklorhydrin
Formel:
ClCH₂CH(O)CH₂ MW: 92,52 g/mol Kokpunkt: 113 - 115 °C (1013 hPa) Smältpunkt: –48 °C Densitet: 1,18 g/cm³ (20 °C) Flampunkt: 34°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00077760 CAS nummer: 67843-74-7 EINECS: 482-080-0 UN: 2023 ADR: 6.1,II |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Specific rotation [a]20/D | +42.0 to +47.0 deg (Neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...