(R)-(-)-Epiklorhydrin ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: (R)-(-)-3-Klor-1,2-epoxipropan, (R)-(-)-1-Klor-2,3-epoxipropan

E0581-25G E0581-5G
TCIAE0581-25GEA 550 SEK
TCIAE0581-25G TCIAE0581-5G
(R)-(-)-Epiklorhydrin ≥98.0%
(R)-(-)-Epiklorhydrin
Formel: ClCH₂CH(O)CH₂
MW: 92,52 g/mol
Kokpunkt: 116°C
Smältpunkt: –48 °C
Densitet: 1,18 g/cm³ (20 °C)
Flampunkt: 31°C
Storage Temperature: Kylskåp
MDL Number: MFCD00077759
CAS nummer: 51594-55-9
EINECS: 424-280-2
UN: 2023
ADR: 6.1,II
Merck Index: 14,03532
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=074403a3e3aee2df9bd44a35f4bc27eae5010c9a&vid=aa6b693d37f853a6cdbb5bd70b1fdf9e3ffce757&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -42.0 to -47.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR