1-(2-Thenoyl)-3,3,3-trifluoroacetone ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: (TTA), Thenoyltrifluoroacetone, 4,4,4-Trifluoro-1-(2-thienyl)butane-1,3-dione, TTFA, 2-Thenoyltrifluoroacetone, (2-Thenoyl)-ω,ω,ω-trifluoroacetone
T0438-25G T0438-500G
TCIAT0438-25GEA 350 SEK
TCIAT0438-25G TCIAT0438-500G
1-(2-Thenoyl)-3,3,3-trifluoroacetone ≥98.0%
1-(2-Thenoyl)-3,3,3-trifluoroacetone
Formel: C₈H₅F₃O₂S
MW: 222,19 g/mol
Kokpunkt: 104°C
Smältpunkt: 40…47 °C
Flampunkt: 111 °C
MDL Number: MFCD00005445
CAS nummer: 326-91-0
EINECS: 206-316-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99f03297e2de3f769bbf91fb03bcda07fa27c33a&vid=99f03297e2de3f769bbf91fb03bcda07fa27c33a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 46.0 deg-C
Solubility in Methanol almost transparency