2,3,4,5,6-Pentafluorostyrene ≥98.0% stabilised

Leverantör: TCI EUROPE
Varning


Stabilization: med 4-tert-Butylpyrokatekol

P0862-25G P0862-5G
TCIAP0862-25GEA 2675 SEK
TCIAP0862-25G TCIAP0862-5G
2,3,4,5,6-Pentafluorostyrene ≥98.0% stabilised
2,3,4,5,6-Pentafluorostyrene
Formel: C₈H₃F₅
MW: 194,1 g/mol
Kokpunkt: 140°C
Densitet: 1,4295 g/cm³ (20 °C)
Flampunkt: 34°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000300
CAS nummer: 653-34-9
EINECS: 211-500-5
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e6fc5b3045e544bd84bb87404fdf6c32d079d11&vid=5e6fc5b3045e544bd84bb87404fdf6c32d079d11&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR