3-Fluorostyrene ≥97.0% stabilised

Leverantör: TCI EUROPE
Varning


Stabilization: med 4-tert-Butylpyrokatekol

F0409-1G F0409-5G
TCIAF0409-1GEA 615 SEK
TCIAF0409-1G TCIAF0409-5G
3-Fluorostyrene ≥97.0% stabilised
3-Fluorostyrene
Formel: C₈H₇F
MW: 122,14 g/mol
Kokpunkt: 41°C
Densitet: 1,025 g/cm³ (20 °C)
Flampunkt: 29°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000341
CAS nummer: 350-51-6
EINECS: 206-504-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d16cdc45b7ce100c2960a6b4b587e9b00251b1d&vid=7d16cdc45b7ce100c2960a6b4b587e9b00251b1d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR