o-Tolylacetonitrile ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
(2-Methylphenyl)acetonitrile, 2-(o-Tolyl)acetonitrile, 2-Methylbenzyl cyanide
X0045-25ML
X0045-100ML
TCIAX0045-25MLEA
710
SEK
TCIAX0045-25ML
TCIAX0045-100ML
o-Tolylacetonitrile ≥98.0%
o-Tolylacetonitrile
Formel:
C₉H₉N MW: 131,18 g/mol Kokpunkt: 120°C Smältpunkt: 58…59 °C Densitet: 1,0156 g/cm³ (22 °C) |
MDL Number:
MFCD00001904 CAS nummer: 22364-68-7 EINECS: 244-937-5 UN: 3276 ADR: 6.1,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...