Butyl vinyl ether ≥98.0% stabilised
Leverantör: TCI
Fara
Synonyms: n-Butyl vinyl ether
Stabilization: Stabilised with potassium hydroxide
B0746-500ML B0746-25ML
TCIAB0746-500MLEA 535 SEK
TCIAB0746-500ML TCIAB0746-25ML
Butyl vinyl ether ≥98.0% stabilised
Butyl vinyl ether
Formel: C₆H₁₂O
Kokpunkt: 93°C
Smältpunkt: -92°C
Densitet: 0,7804 g/cm³ (20 °C)
Flampunkt: -9°C
Storage Temperature: Kylskåp
MDL Number: MFCD00009454
CAS nummer: 111-34-2
EINECS: 203-860-7
UN: 2352
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b5fdfd8267c2c1172c0e7b5f4a18cd4ddaa4e89&vid=3b5fdfd8267c2c1172c0e7b5f4a18cd4ddaa4e89&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %