meso-Tetraphenylporphine ultra-high sensitive spectrophotometric reagent for Cu

Leverantör: TCI EUROPE

Synonyms: 5,10,15,20-Tetraphenyl-21H,23H-porphine, 5,10,15,20-Tetraphenylporphyrin, meso-Tetraphenylporphine, TPP

A5012-1G A5012-25G
TCIAA5012-1GEA 415 SEK
TCIAA5012-1G TCIAA5012-25G
meso-Tetraphenylporphine ultra-high sensitive spectrophotometric reagent for Cu
meso-Tetraphenylporphine
Formel: C₄₄H₃₀N₄
MW: 614,75 g/mol
Densitet: 1,276 g/cm³ (20 °C)
MDL Number: MFCD00011680
CAS nummer: 917-23-7
EINECS: 213-025-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5beb9473f042f6c9ea8a017f4f33fd8b79f90e&vid=958cdff18fe91aa8cd5fe3199fa8df986427f5e8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Dark green to Dark purple to Black powder to crystal
Lambda max. 415.0 to 418.0 nm (DMF)
Molar extinction coefficient min. 420000 (1mg/L, DMF, 415.0 to 418.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR