3-Methyl-1-butanethiol ≥94.0%
Leverantör: TCI
|
Fara
|
Synonyms:
1-Mercapto-3-methylbutane, 3-Methylbutyl mercaptan, Isopentyl mercaptan
I0149-25ML
I0149-250ML
TCIAI0149-25MLEA
485
SEK
TCIAI0149-25ML
TCIAI0149-250ML
3-Methyl-1-butanethiol ≥94.0%
3-Methyl-1-butanethiol
Formel:
C₅H₁₂S MW: 104.22 g/mol Kokpunkt: 120°C Smältpunkt: -134 Flampunkt: 19°C |
MDL Number:
MFCD00004899 CAS nummer: 541-31-1 EINECS: 208-774-3 UN: 3336 ADR: 3 II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 93.0 % |
Purity (Iodometric Titration) | min. 94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...