4-Benzylaniline ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Aminodiphenylmethane

A1260-25G A1260-5G
TCIAA1260-25GEA 3275 SEK
TCIAA1260-25G TCIAA1260-5G
4-Benzylaniline ≥97.0% (by GC, titration analysis)
4-Benzylaniline
Formel: C₁₃H₁₃N
MW: 183,25 g/mol
Kokpunkt: 300 °C (1013 hPa)
Smältpunkt: 35…36 °C
Densitet: 1,07 g/cm³ (20 °C)
MDL Number: MFCD00047861
CAS nummer: 1135-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c083ae586b5959f1e83ff87baa91aadcf59346eb&vid=c083ae586b5959f1e83ff87baa91aadcf59346eb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 36.0 to 43.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR