1,3-Dimethyl-2-thiourea ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1,3-Dimethylthiourea, N,N'-Dimethylthiourea
D0804-25G
D0804-500G
D0804-100G
TCIAD0804-25GEA
590
SEK
TCIAD0804-25G
TCIAD0804-500G
TCIAD0804-100G
1,3-Dimethyl-2-thiourea ≥97.0%
1,3-Dimethyl-2-thiourea
Formel:
C₃H₈N₂S MW: 104,18 g/mol Kokpunkt: 133 °C (1013 hPa) |
MDL Number:
MFCD00004923 CAS nummer: 534-13-4 EINECS: 208-588-2 Merck Index: 14,03261 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Bromination Method) | min. 97.0 % |
Melting point | 64.0 to 66.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...