(S)-(+)-Methyl mandelate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: L(+)-Mandelic acid methyl ester, L(+)-2-Hydroxy-2-phenylacetic acid methyl ester, Methyl (S)-(+)-mandelate, L(+)-Methyl mandelate, (S)-(+)-Mandelic acid methyl ester, (S)-(+)-2-Hydroxy-2-phenylacetic acid methyl ester, L(+)-Methyl glycolate, (S)-(+)-Methyl glycolate

M1350-1G M1350-5G
TCIAM1350-1GEA 245 SEK
TCIAM1350-1G TCIAM1350-5G
(S)-(+)-Methyl mandelate ≥98.0%
(S)-(+)-Methyl mandelate
Formel: C₉H₁₀O₃
MW: 166,18 g/mol
Kokpunkt: 138°C
Smältpunkt: 58 °C
MDL Number: MFCD00064246
CAS nummer: 21210-43-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cd2c0ffe0370228598fe77879e4e1962dfd8b87&vid=7cd2c0ffe0370228598fe77879e4e1962dfd8b87&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 95.0 ee%
Melting point 53.0 to 56.0 deg-C
Specific rotation [a]20/D +138.0 to +142.0 deg (C=1, Methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR