Metyl(R)-(+)-laktat ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Methyl D-(+)-Lactate, (R)-Methyl 2-hydroxypropanoate, D-(+)-Lactic acid methyll ester, Methyl (R)-(+)-2-hydroxypropionate
L0136-25G L0136-5G
TCIAL0136-25GEA 2125 SEK
TCIAL0136-25G TCIAL0136-5G
Metyl(R)-(+)-laktat ≥98.0%
Metyl(R)-(+)-laktat
Formel: C₄H₈O₃
MW: 104,11 g/mol
Kokpunkt: 33°C
Densitet: 1,1037 g/cm³ (11 °C)
Flampunkt: 56°C
MDL Number: MFCD00004517
CAS nummer: 17392-83-5
EINECS: 241-420-6
UN: 3272
ADR: 3,III
Merck Index: 14,06094
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=909ad2f85765c54c4f917d9f9d55f02dddd50e7c&vid=909ad2f85765c54c4f917d9f9d55f02dddd50e7c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 97.0 ee%
Specific rotation [a]20/D +7.9 to +8.5 deg (neat)