2-Bromo-5-methylthiophene ≥98.0% stabilised

Leverantör: TCI

Synonyms: 5-Methyl-2-bromothiophene, 2-Bromo-5-methylthiophenol

Stabilization: Stabilised with copper chip + NaHCO₃

B1540-25G B1540-5G
TCIAB1540-25GEA 2000 SEK
TCIAB1540-25G TCIAB1540-5G
2-Bromo-5-methylthiophene ≥98.0% stabilised
2-Bromo-5-methylthiophene
Formel: C₅H₅BrS
MW: 177.06 g/mol
Kokpunkt: 175°C
Smältpunkt: 58
Flampunkt: 78°C
Storage Temperature: Kylskåp
MDL Number: MFCD00130103
CAS nummer: 765-58-2
EINECS: 212-151-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f89da32c48c2067bebda122bb43d2eaaef1c5cc9&vid=f89da32c48c2067bebda122bb43d2eaaef1c5cc9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR