3,5-Dimethyl-p-anisic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3,5-Dimethyl-p-anisic acid, 4-Methoxy-3,5-dimethylbenzoic acid, 3,5-Dimethyl-4-methoxybenzenecarboxylic acid, 3,5-Dimethyl-4-methoxybenzoic acid
M2139-1G
M2139-5G
TCIAM2139-1GEA
445
SEK
TCIAM2139-1G
TCIAM2139-5G
3,5-Dimethyl-p-anisic acid ≥98.0% (by GC, titration analysis)
3,5-Dimethyl-p-anisic acid
Formel:
C₁₀H₁₂O₃ MW: 180,2 g/mol Kokpunkt: 313 °C (1013 hPa) Smältpunkt: 196…198 °C Densitet: 1,253 g/cm³ (20 °C) |
MDL Number:
MFCD00020309 CAS nummer: 21553-46-8 EINECS: 244-441-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 192.0 to 196.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...