N,N'-Dimethylethylendiamine ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: N1,N2-Dimethylethane-1,2-diamine, sym. Dimethylethylendiamine, N,N'-Dimethylethylenediamine
D0720-25ML D0720-5ML
TCIAD0720-25MLEA 1025 SEK
TCIAD0720-25ML TCIAD0720-5ML
N,N'-Dimethylethylendiamine ≥97.0% (by GC, titration analysis)
N,N'-Dimethylethylendiamine
Formel: C₄H₁₂N₂
MW: 88,15 g/mol
Kokpunkt: 119 °C (1013 hPa)
Densitet: 0,82 g/cm³ (20 °C)
Flampunkt: 26°C
MDL Number: MFCD00008290
CAS nummer: 110-70-3
EINECS: 203-793-3
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ce2bfb138a409ad0829e55dad61c325d2c43ea1&vid=2ce2bfb138a409ad0829e55dad61c325d2c43ea1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %