N,N-Dimethylformamide dipropyl acetal ≥90.0% for esterification

Leverantör: TCI
Varning

Synonyms: 1,1-Dipropoxytrimethylamine, N,n-dimethylformamidedipropylacetal, 1,1-Dipropoxy-N,N-dimethylmethylamine

D1301-25ML D1301-5ML
TCIAD1301-25MLEA 950 SEK
TCIAD1301-25ML TCIAD1301-5ML
N,N-Dimethylformamide dipropyl acetal ≥90.0% for esterification
N,N-Dimethylformamide dipropyl acetal
Formel: C₉H₂₁NO₂
Kokpunkt: 67 °C (1013 hPa)
Densitet: 0,8537 g/cm³ (20 °C)
Flampunkt: 46°C
MDL Number: MFCD00009374
CAS nummer: 6006-65-1
EINECS: 227-855-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebb7c90eb931a630ca5240617e4c8e910e2202a8&vid=07029c25c3533e7ee65f96860e27497a378cff87&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (Nonaqueous Titration) min. 90.0 %
Effect test of GC derivalization Effective as derivatization agent Myristic Acid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR