3'-Methylacetanilide ≥98.0%
Leverantör: TCI
Varning
A0062-25G A0062-250G
TCIAA0062-25GEA 565 SEK
TCIAA0062-25G TCIAA0062-250G
3'-Methylacetanilide ≥98.0%
3'-Methylacetanilide
Formel: C₉H₁₁NO
Kokpunkt: 163°C
Smältpunkt: 62…63 °C
Densitet: 1,141 g/cm³ (15 °C)
MDL Number: MFCD00014962
CAS nummer: 537-92-8
EINECS: 208-678-1
Merck Index: 14,00074
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3a8399eb01c7b7846d36c149883d422832f059e&vid=c3a8399eb01c7b7846d36c149883d422832f059e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C
Solubility in Methanol almost transparency