3-Methyl-1-cyclohexene ≥93.0%

Leverantör: TCI
Fara

M1034-25ML M1034-500ML
TCIAM1034-25MLEA 685 SEK
TCIAM1034-25ML TCIAM1034-500ML
3-Methyl-1-cyclohexene ≥93.0%
3-Methyl-1-cyclohexene
Formel: C₇H₁₂
Kokpunkt: 105°C
Smältpunkt: –124 °C
Densitet: 0,805 g/cm³ (20 °C)
Flampunkt: –3°C
MDL Number: MFCD00001571
CAS nummer: 591-48-0
EINECS: 209-717-5
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d54a652add3c4f169c51afa9855710f6a7fb2a17&vid=d54a652add3c4f169c51afa9855710f6a7fb2a17&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR