3-Hydroxyphenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

Synonyms: (3-Hydroxyphenyl)metaboric acid, (3-Hydroxyphenyl)boronic acid, Dihydroxy(3-hydroxyphenyl)borane, 3-Hydroxybenzeneboronic acid

H1185-1G H1185-5G
TCIAH1185-1GEA 415 SEK
TCIAH1185-1G TCIAH1185-5G
3-Hydroxyphenylboronic acid (contains varying amounts of Anhydride)
3-Hydroxyphenylboronic acid
Formel: C₆H₇BO₃
MW: 137,93 g/mol
MDL Number: MFCD01074603
CAS nummer: 87199-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f8cb02c8a57f70ddfc40345d61f4692cf939125&vid=0f8cb02c8a57f70ddfc40345d61f4692cf939125&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (Neutralization titration) 97.0 to 115.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR