Bis(2-ethylhexyl)amine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: Dioctylamine, Di(2-ethylhexyl)amine, Bis(2-ethylhexyl)amine
D0510-25ML D0510-500ML
TCIAD0510-25MLEA 215 SEK
TCIAD0510-25ML TCIAD0510-500ML
Bis(2-ethylhexyl)amine ≥98.0% (by GC, titration analysis)
Bis(2-ethylhexyl)amine
Formel: C₁₆H₃₅N
MW: 241,46 g/mol
Kokpunkt: 281,1 °C (1013 hPa)
Smältpunkt: < –60 °C
Densitet: 0,8062 g/cm³ (20 °C)
Flampunkt: 121°C
Storage Temperature: Omgivande
MDL Number: MFCD00009489
CAS nummer: 106-20-7
EINECS: 203-372-4
UN: 2922
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24eb1ed4617a26fbb4a59f143c4b8b6df933cd38&vid=24eb1ed4617a26fbb4a59f143c4b8b6df933cd38&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %