Fenolftalein ACS
Leverantör: VWR Chemicals
Fara
Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
0376-100GEA 1200 SEK
0376-100G 0376-50G
Fenolftalein ACS
Fenolftalein
Formel: C₂₀H₁₄O₄
MW: 318.33 g/mol
Smältpunkt: 258…262 °C
Densitet: 1.3 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00005913
CAS nummer: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Clarity of Alcohol Solution Pass
Visual Transition Interval (pH 8 – 10) Pass