3-Hydroxypropiononitrile 95,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: Ethylene Cyanohydrin, 2-Cyanoethanol, 3-Hydroxypropionitrile, Hydracrylonitrile, β-Hydroxypropiononitrile

C1285-25ML C1285-500ML
TCIAC1285-25MLEA 405 SEK
TCIAC1285-25ML TCIAC1285-500ML
3-Hydroxypropiononitrile 95,0 GC_ANALYSMETOD
3-Hydroxypropiononitrile
Formel: C₃H₅NO
MW: 71.08 g/mol
Kokpunkt: 221°C
Smältpunkt: -46
Flampunkt: 137°C
Storage Temperature: Omgivande
MDL Number: MFCD00002826
CAS nummer: 109-78-4
EINECS: 203-704-8
Merck Index: 14,03794
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c994066873bf6895548669ce9edfe61f16f51399&vid=c994066873bf6895548669ce9edfe61f16f51399&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR