4-(1-Piperidinyl)aniline

Leverantör: Thermo Scientific

Synonyms: 4-Piperidinoaniline

BTB05830DA BTB05830EA
MAYBBTB05830DAEA 0 SEK
MAYBBTB05830DA MAYBBTB05830EA
4-(1-Piperidinyl)aniline
4-(1-Piperidinyl)aniline
Formel: C₁₁H₁₆N₂
MW: 176,26 g/mol
MDL Number: MFCD00051688
CAS nummer: 2359-60-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR