4-Benzyl-4-hydroxypiperidine ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Benzylpiperidin-4-ol
A15664.03 A15664.06 A15664.14
A15664.03EA 1025 SEK
A15664.03 A15664.06 A15664.14
4-Benzyl-4-hydroxypiperidine ≥97%
4-Benzyl-4-hydroxypiperidine
Purity: 97% CAS No.: 51135-96-7 Molecular Formula: C12H17NO Formula Weight: 191.27 Melting Point: ca 80° Beilstein Reference: 1451803
Formel: C₁₂H₁₇NO
MW: 191,27 g/mol
Smältpunkt: ∼ 80 °C
Storage Temperature: Omgivande
MDL Number: MFCD00023147
CAS nummer: 51135-96-7
EINECS: 257-003-7